Gene Transcripts

Gene Symbol
LOC100911069
Entrez Gene ID
100911069
Gene Name
protein FAM54A-like
Gene Aliases
Dufd1, Mtfr2
Chromosome Location
Chr. 1: 15834779 - 15850453 on Build Rnor_6.0
UniGene ID
Rn.202401
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290110.1 NP_001277039.1 2, 3 208
XM_006227672.2 XP_006227734.1 1, 2 276
GenBank mRNA BC159435.1 - 2, 3 208

View Full Product Details