Gene Transcripts

Gene Symbol
Nepn
Entrez Gene ID
309775
Gene Name
nephrocan
Gene Aliases
RGD1307755
Chromosome Location
Chr. 20: 33527932 - 33543860 on Build Rnor_6.0
UniGene ID
Rn.42078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107632.1 NP_001101102.1 3 1185
XM_006256491.2 XP_006256553.1 3 1470

View Full Product Details