Gene Transcripts

Gene Symbol
Arse
Entrez Gene ID
310326
Gene Name
arylsulfatase E
Gene Aliases
Chromosome Location
Chr. 2: 122876645 - 122884673 on Build Rnor_6.0
UniGene ID
Rn.79118
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047885.1 NP_001041350.1 6 925
XM_006232280.2 XP_006232342.1 7 1112
XM_006232281.2 XP_006232343.1 7 1117
GenBank mRNA BC168644.1 - 5 2118

View Full Product Details