Gene Transcripts

Gene Symbol
Pla2g4b
Entrez Gene ID
311341
Gene Name
phospholipase A2, group IVB (cytosolic)
Gene Aliases
Chromosome Location
Chr. 3: 111825004 - 111833526 on Build Rnor_6.0
UniGene ID
Rn.21662
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107764.2 NP_001101234.2 13 1288
XM_006234775.1 XP_006234837.1 12 1220
XM_008762103.1 XP_008760325.1 12 1861

View Full Product Details