Gene Transcripts

Gene Symbol
Cbx6
Entrez Gene ID
315136
Gene Name
chromobox homolog 6
Gene Aliases
Chromosome Location
Chr. 7: 121047996 - 121058142 on Build Rnor_6.0
UniGene ID
Rn.161995
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012119.1 NP_001012119.1 5 1054
XM_006241994.2 XP_006242056.1 5 1221
GenBank mRNA BC087122.1 - 5 1054

View Full Product Details