Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001007557.1 | NP_001007558.1 | 9 | 1011 | |
XM_006244280.2 | XP_006244342.1 | 9 | 1032 | ||
XM_008766759.1 | XP_008764981.1 | 8 | 1067 | ||
GenBank mRNA | AY686632.1 | - | 9 | 1011 |