Gene Transcripts

Gene Symbol
Haus5
Entrez Gene ID
100362495
Gene Name
HAUS augmin-like complex, subunit 5
Gene Aliases
Chromosome Location
Chr. 1: 89113046 - 89124899 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_002728731.3 XP_002728777.1 8, 9 759
XM_008759256.1 XP_008757478.1 8, 9 751

View Full Product Details