Gene Transcripts

Gene Symbol
Plxnb3
Entrez Gene ID
363517
Gene Name
plexin B3
Gene Aliases
RGD1560615
Chromosome Location
Chr. X: 157015297 - 157030152 on Build Rnor_6.0
UniGene ID
Rn.101173
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135878.1 NP_001129350.1 23 4258
XM_006229587.2 XP_006229649.1 24 4416
XM_008773630.1 XP_008771852.1 24 4416
XM_008773631.1 XP_008771853.1 25 4623
XM_008773632.1 XP_008771854.1 23 4254

View Full Product Details