Gene Transcripts

Gene Symbol
Kdm6b
Entrez Gene ID
363630
Gene Name
lysine (K)-specific demethylase 6B
Gene Aliases
Jmjd3
Chromosome Location
Chr. 10: 56001626 - 56009582 on Build Rnor_6.0
UniGene ID
Rn.205533
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108829.1 NP_001102299.1 4 388
GenBank mRNA EF532386.1 - 847
EF532387.1 - 845
EF532388.1 - 847
EF532389.1 - 846

View Full Product Details