Gene Transcripts

Gene Symbol
Ces2g
Entrez Gene ID
291952
Gene Name
carboxylesterase 2G
Gene Aliases
Ces2l, RGD1308358
Chromosome Location
Chr. 19: 37025379 - 37034611 on Build Rnor_6.0
UniGene ID
Rn.151252
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106175.1 NP_001099645.1 7 1067
XM_006255448.2 XP_006255510.1 6 815

View Full Product Details