Gene Transcripts

Gene Symbol
RT1-M4
Entrez Gene ID
309584
Gene Name
RT1 class Ib, locus M4
Gene Aliases
RT1.M4
Chromosome Location
Chr. 20: 2043521 - 2048162 on Build Rnor_6.0
UniGene ID
Rn.222433
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168343.1 NP_001161815.1 3 570
XM_006255871.2 XP_006255933.1 3 768

View Full Product Details