Gene Transcripts

Gene Symbol
Chadl
Entrez Gene ID
100910434
Gene Name
chondroadherin-like
Gene Aliases
Chromosome Location
Chr. 7: 122915847 - 122929114 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008765807.1 XP_008764029.1 2 609
XM_008765808.1 XP_008764030.1 2 609
XM_008765809.1 XP_008764031.1 3 1949

View Full Product Details