Gene Transcripts

Gene Symbol
Crtc3
Entrez Gene ID
365297
Gene Name
CREB regulated transcription coactivator 3
Gene Aliases
RGD1309666
Chromosome Location
Chr. 1: 142398248 - 142500558 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006229462.2 XP_006229524.1 11 1189
XM_006229463.2 XP_006229525.1 11 1189

View Full Product Details