Gene Transcripts

Gene Symbol
Ifitm7
Entrez Gene ID
288244
Gene Name
interferon induced transmembrane protein 7
Gene Aliases
Chromosome Location
Chr. 11: 34037103 - 34037519 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_221637.1 XP_221637.1 1 386

View Full Product Details