Gene Transcripts

Gene Symbol
Resp18
Entrez Gene ID
50561
Gene Name
regulated endocrine-specific protein 18
Gene Aliases
Chromosome Location
Chr. 9: 82470794 - 82477136 on Build Rnor_6.0
UniGene ID
Rn.66430
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019278.1 NP_062151.1 3 319
GenBank mRNA BC058147.1 - 3 315
L25633.1 - 3 319

View Full Product Details