Gene Transcripts

Gene Symbol
Cox8b
Entrez Gene ID
25250
Gene Name
cytochrome c oxidase, subunit VIIIb
Gene Aliases
CCO8A, COXVIII-H, Cox8h
Chromosome Location
Chr. 1: 213648787 - 213650247 on Build Rnor_6.0
UniGene ID
Rn.10325
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012786.3 NP_036918.2 1 108
GenBank mRNA BF411029.1 - 2 226
FM054348.1 - 1 108
X64827.1 - 1 103

View Full Product Details