Gene Transcripts

Gene Symbol
Psat1
Entrez Gene ID
293820
Gene Name
phosphoserine aminotransferase 1
Gene Aliases
Psa1
Chromosome Location
Chr. 1: 233124089 - 233145941 on Build Rnor_6.0
UniGene ID
Rn.100813
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198738.2 NP_942033.2 9 1090
GenBank mRNA AY239016.1 - 9 1078
BC079352.1 - 9 1090 7108218

View Full Product Details