Gene Transcripts

Gene Symbol
Banf2
Entrez Gene ID
296198
Gene Name
barrier to autointegration factor 2
Gene Aliases
RGD1307909
Chromosome Location
Chr. 3: 138128131 - 138183750 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235132.2 XP_006235194.1 5 619
XM_006235134.2 XP_006235196.1 7 1524
XM_006235135.2 XP_006235197.1 5 1313
XM_006235137.2 XP_006235199.1 4 461
XM_008762281.1 XP_008760503.1 4 583
XM_008762282.1 XP_008760504.1 5 1269
XM_008762283.1 XP_008760505.1 3 300
XM_215865.7 XP_215865.3 4 1194

View Full Product Details