Gene Transcripts

Gene Symbol
Enpp7
Entrez Gene ID
303729
Gene Name
ectonucleotide pyrophosphatase/phosphodiesterase 7
Gene Aliases
E-NPP 7
Chromosome Location
Chr. 10: 108095131 - 108099483 on Build Rnor_6.0
UniGene ID
Rn.211790
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012466.1 NP_001012484.1 3 599
GenBank mRNA AY568760.1 - 3 599

View Full Product Details