Gene Transcripts

Gene Symbol
Epc1
Entrez Gene ID
100362678
Gene Name
enhancer of polycomb homolog 1 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 17: 57303968 - 57395004 on Build Rnor_6.0
UniGene ID
Rn.219240
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC098858.1 - 1 360

View Full Product Details