Gene Transcripts

Gene Symbol
Igfl3
Entrez Gene ID
361516
Gene Name
IGF-like family member 3
Gene Aliases
Ceacam12
Chromosome Location
Chr. 1: 79474809 - 79481865 on Build Rnor_6.0
UniGene ID
Rn.17980
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108474.1 NP_001101944.1 2 476
GenBank mRNA BC099083.1 - 2 435
BC158846.1 - 2 461 7443540

View Full Product Details