Gene Transcripts

Gene Symbol
Ostc
Entrez Gene ID
362040
Gene Name
oligosaccharyltransferase complex subunit (non-catalytic)
Gene Aliases
Dc2, RGD1560708
Chromosome Location
Chr. 2: 235816328 - 235831651 on Build Rnor_6.0
UniGene ID
Rn.1889
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108566.1 NP_001102036.1 3 338
GenBank mRNA BC159425.1 - 3 324
FQ219812.1 - 3 344
FQ219920.1 - 3 352
FQ220255.1 - 3 320
FQ230162.1 - 3 320
FQ234814.1 - 3 300

View Full Product Details