Gene Transcripts

Gene Symbol
Ccdc88c
Entrez Gene ID
362770
Gene Name
coiled-coil domain containing 88C
Gene Aliases
RGD1307429
Chromosome Location
Chr. 6: 124905811 - 125028013 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_343096.6 XP_343097.4 3 693

View Full Product Details