Gene Transcripts

Gene Symbol
Wdr38
Entrez Gene ID
366035
Gene Name
WD repeat domain 38
Gene Aliases
RGD1562587
Chromosome Location
Chr. 3: 23290810 - 23294818 on Build Rnor_6.0
UniGene ID
Rn.198114
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_575125.5 XP_575125.6 1 102
GenBank mRNA FQ213095.1 - 1 96

View Full Product Details