Gene Transcripts

Gene Symbol
Krt42
Entrez Gene ID
450231
Gene Name
keratin 42
Gene Aliases
Ka22
Chromosome Location
Chr. 10: 88165667 - 88172910 on Build Rnor_6.0
UniGene ID
Rn.142051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008816.1 NP_001008816.1 1 313

View Full Product Details