Gene Transcripts

Gene Symbol
Klra2
Entrez Gene ID
494193
Gene Name
killer cell lectin-like receptor, subfamily A, member 2
Gene Aliases
Ly49i6
Chromosome Location
Chr. 4: 165005722 - 165026566 on Build Rnor_6.0
UniGene ID
Rn.134170
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009718.1 NP_001009718.1 2 167
XM_008763403.1 XP_008761625.1 2 319
GenBank mRNA AY651018.1 - 2 167

View Full Product Details