Gene Transcripts

Gene Symbol
Naa30
Entrez Gene ID
498489
Gene Name
N(alpha)-acetyltransferase 30, NatC catalytic subunit
Gene Aliases
Nat12, RGD1559923
Chromosome Location
Chr. 15: 26032758 - 26052774 on Build Rnor_6.0
UniGene ID
Rn.15205
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109099.1 NP_001102569.1 4 1034
XM_006251813.2 XP_006251875.1 5 1821

View Full Product Details