Gene Transcripts

Gene Symbol
Rgs9bp
Entrez Gene ID
499132
Gene Name
regulator of G protein signaling 9 binding protein
Gene Aliases
RGD1561747
Chromosome Location
Chr. 1: 91855270 - 91855983 on Build Rnor_6.0
UniGene ID
Rn.218592
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109142.1 NP_001102612.1 1 57

View Full Product Details