Gene Transcripts

Gene Symbol
RGD1563348
Entrez Gene ID
502642
Gene Name
similar to Selenoprotein H
Gene Aliases
Selh
Chromosome Location
Chr. 3: 72070384 - 72071895 on Build Rnor_6.0
UniGene ID
Rn.63
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114939.1 NP_001108411.1 3 377
GenBank mRNA AI030980.1 - 2 273
BI285703.1 - 2 295
FQ220990.1 - 3 386

View Full Product Details