Gene Transcripts

Gene Symbol
Dbx2
Entrez Gene ID
541457
Gene Name
developing brain homeobox 2
Gene Aliases
Chromosome Location
Chr. 7: 136966879 - 136997104 on Build Rnor_6.0
UniGene ID
Rn.93216
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001053826.4 XP_001053826.1 3 837
XM_006242286.2 XP_006242348.1 2 741
GenBank mRNA AB121147.1 - 1 143

View Full Product Details