Gene Transcripts

Gene Symbol
Mcpt4
Entrez Gene ID
54270
Gene Name
mast cell protease 4
Gene Aliases
Chromosome Location
Chr. 15: 34751550 - 34754409 on Build Rnor_6.0
UniGene ID
Rn.10698
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019321.2 NP_062194.1 4 541
GenBank mRNA U67907.1 - 4 543

View Full Product Details