Gene Transcripts

Gene Symbol
Ppapdc2
Entrez Gene ID
619549
Gene Name
phosphatidic acid phosphatase type 2 domain containing 2
Gene Aliases
Chromosome Location
Chr. 1: 247084228 - 247087080 on Build Rnor_6.0
UniGene ID
Rn.7335
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034854.1 NP_001030026.1 1 826
GenBank mRNA BC081970.1 - 1 826

View Full Product Details