Gene Transcripts

Gene Symbol
Vom2r7
Entrez Gene ID
685557
Gene Name
vomeronasal 2 receptor, 7
Gene Aliases
Chromosome Location
Chr. 1: 54616669 - 54632656 on Build Rnor_6.0
UniGene ID
Rn.217943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099463.1 NP_001092933.1 127

View Full Product Details