Gene Transcripts
- Gene Symbol
- Klk1b7-ps
- Entrez Gene ID
- 16604
- Gene Name
- kallikrein 1-related peptidase b7, pseudogene
- Gene Aliases
- 1700127D06Rik, Klk-ps2, mGK-7
- Chromosome Location
- Chr. 7: 43945012 - 43945927 on Build GRCm38
- UniGene ID
- Mm.195787
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_033120.1 |
|
3 |
308 |
|
GenBank mRNA |
AK007298.1 |
- |
|
310 |
|
View Full Product Details