Gene Transcripts
- Gene Symbol
- Speer9-ps1
- Entrez Gene ID
- 73949
- Gene Name
- spermatogenesis associated glutamate (E)-rich protein 9, pseudogene 1
- Gene Aliases
- 4930415A14Rik, SPEER-9, Speer9-ps
- Chromosome Location
- Chr. 7: 3128146 - 3144992 on Build GRCm38
- UniGene ID
- Mm.55687
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_001583.3 |
|
3 |
332 |
|
GenBank mRNA |
BY714690.1 |
- |
3 |
333 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK015142.1 |
- |
|
333 |
|
Chr. - - [-] |
View Full Product Details