Gene Transcripts

Gene Symbol
Speer9-ps1
Entrez Gene ID
73949
Gene Name
spermatogenesis associated glutamate (E)-rich protein 9, pseudogene 1
Gene Aliases
4930415A14Rik, SPEER-9, Speer9-ps
Chromosome Location
Chr. 7: 3128146 - 3144992 on Build GRCm38
UniGene ID
Mm.55687
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_001583.3 3 424
GenBank mRNA BY714690.1 - 3 425


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK015142.1 - 425 Chr. - - [-]

View Full Product Details