Gene Transcripts

Gene Symbol
Kis2
Entrez Gene ID
751866
Gene Name
Kaplan integration site 2
Gene Aliases
Kis2-T1, Kis2-T2, Kis2-T3, Kis2-T4, Kis2-T5, Xpcl1
Chromosome Location
Chr. X: 52742563 - 52744593 on Build GRCm38
UniGene ID
Mm.381916
Species
Mus musculus
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003188.1
1 890
GenBank mRNA AK084356.1 -
2 322
AY940614.1 - LIT3387
Kis2
1 890
AY940615.1 - LIT3388
Kis2
2 576
AY940616.1 - LIT3389
Kis2
1 209

View Full Product Details