Gene Transcripts
- Gene Symbol
- Asb7
- Entrez Gene ID
- 117589
- Gene Name
- ankyrin repeat and SOCS box-containing 7
- Gene Aliases
- AI449039, Asb-7, D030055C23Rik
- Chromosome Location
- Chr. 7: 66644565 - 66689650 on Build GRCm38
- UniGene ID
- Mm.34785
- Species
- Mus musculus
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN526221.1 |
- |
1, 2 |
277 |
|
Chr. 7 - 66659893 - 66679350 [-] |
GenBankmRNA |
CK635170.1 |
- |
1, 2 |
111 |
|
Chr. 7 - 66660111 - 66679185 [-] |
GenBankmRNA |
CF534420.1 |
- |
1, 2 |
160 |
|
Chr. 7 - 66659728 - 66679234 [-] |
GenBankmRNA |
CF534183.1 |
- |
2, 3 |
363 |
|
Chr. 7 - 66659964 - 66681799 [-] |
GenBankmRNA |
CK639788.1 |
- |
1, 2 |
277 |
|
Chr. 7 - 66659900 - 66679350 [-] |
View Full Product Details