Gene Transcripts

Gene Symbol
Vps39
Entrez Gene ID
269338
Gene Name
vacuolar protein sorting 39 (yeast)
Gene Aliases
A230065P22Rik, AW743070, Vam6, Vam6P, mVam6
Chromosome Location
Chr. 2: 120316461 - 120353144 on Build GRCm38
UniGene ID
Mm.246131
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027618.1 3 317
GenBank mRNA AK170831.1 - 3 307
BB655861.1 - 3 273

View Full Product Details