Gene Transcripts

Gene Symbol
Psg-ps1
Entrez Gene ID
232919
Gene Name
pregnancy specific glycoprotein pseudogene 1
Gene Aliases
AV036018, Cea-6, Cea6
Chromosome Location
Chr. 7: 17672313 - 17682060 on Build GRCm38
UniGene ID
Mm.153893
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002857.1 4 1256
GenBank mRNA AK132281.1 - 4 1256
M83348.1 - 4 1252

View Full Product Details