Gene Transcripts
- Gene Symbol
- Zf12
- Entrez Gene ID
- 331360
- Gene Name
- Zf12 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. X|NT_165789: 268799 - 270075 on Build GRCm38
- UniGene ID
- Mm.490244
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003547.2 |
|
1 |
1184 |
|
GenBank mRNA |
AK084191.1 |
- |
1 |
1190 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB466068.2 |
- |
1 |
566 |
|
Chr. X|NT_165789.2 - 269417 - 270075 [+] |
View Full Product Details