Gene Transcripts
- Gene Symbol
- AKAP17A
- Entrez Gene ID
- 8227
- Gene Name
- A-kinase anchoring protein 17A
- Gene Aliases
- 721P, AKAP-17A, CCDC133, CXYorf3, DXYS155E, PRKA17A, SFRS17A, XE7, XE7Y
- Chromosome Location
- Chr. Y: 1591593 - 1602520 on Build GRCh38
- UniGene ID
- Hs.522572
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027383.1 |
|
5 |
1408 |
|
GenBank mRNA |
BC028151.1 |
- |
4 |
2081 |
|
L03426.1 |
- |
5 |
1378 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL045946.1 |
- |
2 |
90 |
|
Chr. X - 1599403 - 1601063 [+] |
View Full Product Details