Gene Transcripts

Gene Symbol
NHEG1
Entrez Gene ID
100294720
Gene Name
neuroblastoma highly expressed 1
Gene Aliases
Chromosome Location
Chr. 6: 136982159 - 136993231 on Build GRCh38
UniGene ID
Hs.520412
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_027994.1 3 1163
GenBank mRNA BC025319.1 - 3 1168
BC033111.1 - 3 1168
BC080649.1 - 3 1168

View Full Product Details