Gene Transcripts
- Gene Symbol
- CTAGE11P
- Entrez Gene ID
- 647288
- Gene Name
- CTAGE family member 11, pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 13: 75237753 - 75240381 on Build GRCh38
- UniGene ID
- Hs.567920
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027466.1 |
|
1 |
2039 |
|
GenBank mRNA |
AL133018.1 |
- |
1 |
2039 |
|
View Full Product Details