Gene Transcripts
- Gene Symbol
- CROCCP3
- Entrez Gene ID
- 114819
- Gene Name
- ciliary rootlet coiled-coil, rootletin pseudogene 3
- Gene Aliases
- CROCCL2, dJ798A10.2
- Chromosome Location
- Chr. 1: 16467436 - 16492701 on Build GRCh38
- UniGene ID
- Hs.597881
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023386.1 |
|
15 |
2915 |
|
GenBank mRNA |
AB067509.1 |
- |
15 |
2914 |
|
View Full Product Details