Gene Transcripts

Gene Symbol
DPY19L2P4
Entrez Gene ID
442523
Gene Name
DPY19L2 pseudogene 4
Gene Aliases
Chromosome Location
Chr. 7: 90119400 - 90125600 on Build GRCh38
UniGene ID
Hs.406964
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003551.1 3 701
GenBank mRNA BC031277.1 - 3 658
BC047471.1 - 3 649
BX537815.1 - 3 701
DQ287934.1 - 3 701

View Full Product Details