Gene Transcripts
- Gene Symbol
- ANXA2P2
- Entrez Gene ID
- 304
- Gene Name
- annexin A2 pseudogene 2
- Gene Aliases
- ANX2L2, ANX2P2, LPC2B
- Chromosome Location
- Chr. 9: 33624225 - 33625534 on Build GRCh38
- UniGene ID
- Hs.534301
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003573.1 |
|
1 |
260 |
|
View Full Product Details