Gene Transcripts
- Gene Symbol
- DBIL5P
- Entrez Gene ID
- 100131454
- Gene Name
- diazepam binding inhibitor-like 5, pseudogene
- Gene Aliases
- ELP2P
- Chromosome Location
- Chr. 17: 752333 - 755336 on Build GRCh38
- UniGene ID
- Hs.662289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024120.2 |
|
1 |
1254 |
|
GenBank mRNA |
AF229804.1 |
- |
1 |
927 |
|
View Full Product Details