Gene Transcripts
- Gene Symbol
- RPL21P44
- Entrez Gene ID
- 402176
- Gene Name
- ribosomal protein L21 pseudogene 44
- Gene Aliases
- RPL21_17_477
- Chromosome Location
- Chr. 4: 53985486 - 53987282 on Build GRCh38
- UniGene ID
- Hs.452943
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_027153.1 |
|
1 |
822 |
|
GenBank mRNA |
BC057822.1 |
- |
2 |
793 |
|
View Full Product Details