Gene Transcripts
- Gene Symbol
- TMEM67
- Entrez Gene ID
- 91147
- Gene Name
- transmembrane protein 67
- Gene Aliases
- JBTS6, MECKELIN, MKS3, NPHP11, TNEM67
- Chromosome Location
- Chr. 8: 93754844 - 93832653 on Build GRCh38
- UniGene ID
- Hs.116240
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_024522.1 |
|
29 |
3174 |
|
GenBank mRNA |
BC032835.2 |
- |
29 |
3092 |
|
View Full Product Details